Description
Increase your application capabilities with a variety of accessories for the Thermo Scientific™ Compact Dry Baths/Block Heaters.
Specifications
Specifications
| Description | Heat Insulator for 4 block |
| For Use With (Equipment) | Thermo Scientific™ Compact Dry Bath/Block Heater |
| Lighting | LED |
| Type | Heat Insulator |
| Dimensions (L x W x H) | 36 x 108 x 72 mm |