Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Compact Ear Plug Dispenser

Catalog No. 19104415 Shop All Brady Corporation Products
Change view
Click to view available options
Product Type:
With cover
Without cover
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-104-415 With cover
19-104-414 Without cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-104-415 Supplier Brady™ Supplier No. 2019L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acrylic compact dispensers serve as a visual reminder for employees to wear their ear plugs. Conveniently mount near hazardous work areas to encourage compliance.

  • Clear acrylic with green HEARING PROTECTION text legend
  • Can be wall or table mounted for convenient access
  • Size is 3 X 9 in. with a 6 in. back plate
  • Available with or without top cover
  • Ear plugs not included

Specifications

Depth (English) 6 in.
Depth (Metric) 15.2 cm
For Use With (Equipment) Ear plugs
For Use With (Application) Ear Plug Dispenser with Visual Reminder for Employees to Wear Ear Plugs When Mounted Near Hazardous Work Areas.
Height (English) 3 in.
Height (Metric) 7.6 cm
Holds Ear plugs
Width (Metric) 22.8 cm
Width (English) 9 in.
Product Type With cover
Disposable Reusable
Material Acrylic
Mounting Wall Mountable
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.