Learn More
METTLER TOLEDO 3P INDUSTRIAL Compact Ionizer USB with adapter
Catalog No. NC1711399
Encompass_Preferred
Built-in compact antistatic kit for XPR analytical and micro balances. The compact antistatic kit eliminates positive and negative charges from the container and sample. The ionizer fixes onto the balance and does not need an external transformer.
Catalog No. NC1711399 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 30499860
Add to Cart
Add to Cart
Request a Quote
Built-in compact antistatic kit for XPR analytical and micro balances. The compact antistatic kit eliminates positive and negative charges from the container and sample. The ionizer fixes onto the balance and does not need an external transformer.