Learn More
BURKLE INC Compact jerrycan, conductive, without threaded connector 1.32 gal. (5l)
Catalog No. 5000025835
Encompass_Preferred
The space-saving canister made of electrically conductive HDPE is specially designed for use in explosion-protected areas. When properly earthed, the risk of ignition due to electrostatic charging is avoided, suitable for use in hazardous areas.
Catalog No. 50-000-25835 Supplier BURKLE INC Supplier No. 04351055
The space-saving canister made of electrically conductive HDPE is specially designed for use in explosion-protected areas. When properly earthed, the risk of ignition due to electrostatic charging is avoided, suitable for use in hazardous areas.