Learn More
BURKLE INC Compact jerrycan, conductive, with thr.connector 1.32 gal. (5l)
Supplier:  BURKLE INC 04352055
Encompass_Preferred
The space-saving canister with 3/4" thread not only enables space-saving storage but also convenient handling of the liquid. The space-saving canister made of electrically conductive HDPE is specially designed for use in explosion-protected areas.
Catalog No. 50-000-25795