Learn More
BURKLE INC Compact jerrycan, conductive, with thr.connector 2.64 gal. (10l)
Catalog No. 5000025796
Encompass_Preferred
The space-saving canister with 3/4" thread not only enables space-saving storage but also convenient handling of the liquid. The space-saving canister made of electrically conductive HDPE is specially designed for use in explosion-protected areas.
Catalog No. 50-000-25796 Supplier BURKLE INC Supplier No. 04352105
The space-saving canister with 3/4" thread not only enables space-saving storage but also convenient handling of the liquid. The space-saving canister made of electrically conductive HDPE is specially designed for use in explosion-protected areas.