Learn More
BURKLE INC Compact jerrycan, with threaded connector 1.32 gal. (5l)
Catalog No. NC2187652
Encompass_Preferred
The space-saving canister with 3/4" threaded connection not only saves storage space, but also allows convenient handling of the liquid, e.g. direct filling of liquids. The flat canister can be fitted with a drain tap or other suitable accessories.
This item is not returnable. View return policy
Catalog No. NC2187652 Supplier BURKLE INC Supplier No. 04352050
The space-saving canister with 3/4" threaded connection not only saves storage space, but also allows convenient handling of the liquid, e.g. direct filling of liquids. The flat canister can be fitted with a drain tap or other suitable accessories.