Learn More
BURKLE INC Compact jerrycan, conductive, without threaded connector 2.64 gal. (10l)
Catalog No. 5000025820
Encompass_Preferred
The space-saving canister made of electrically conductive HDPE is specially designed for use in explosion-protected areas. When properly earthed, the risk of ignition due to electrostatic charging is avoided, suitable for use in hazardous areas.
Catalog No. 50-000-25820 Supplier BURKLE INC Supplier No. 04351105
The space-saving canister made of electrically conductive HDPE is specially designed for use in explosion-protected areas. When properly earthed, the risk of ignition due to electrostatic charging is avoided, suitable for use in hazardous areas.