Learn More
BURKLE INC Compact jerrycan, with thread. connector 2.64 gal. (10l)
Catalog No. 5000025977
Encompass_Preferred
The space-saving canister with 3/4" threaded connection not only saves storage space, but also allows convenient handling of the liquid, e.g. direct filling of liquids. The flat canister can be fitted with a drain tap or other suitable accessories.
Catalog No. 50-000-25977 Supplier BURKLE INC Supplier No. 04352100
The space-saving canister with 3/4" threaded connection not only saves storage space, but also allows convenient handling of the liquid, e.g. direct filling of liquids. The flat canister can be fitted with a drain tap or other suitable accessories.