Learn More
BURKLE INC Compact jerrycan & StopCock set 2.64 gal. (10l)
Catalog No. 5000025941
Encompass_Preferred
The StopCock space-saving kit, consisting of the space-saving canister with 3/4" threaded connection and the StopCock drain tap, enables space-saving storage as well as convenient handling of the liquid through direct filling or clarification.
Catalog No. 50-000-25941 Supplier BURKLE INC Supplier No. 04350010
The StopCock space-saving kit, consisting of the space-saving canister with 3/4" threaded connection and the StopCock drain tap, enables space-saving storage as well as convenient handling of the liquid through direct filling or clarification.