Learn More
BURKLE INC Compact jerrycan & StopCock set, 2.64 gal. (10l), conductive
Catalog No. 5000025738
Encompass_Preferred
The electrically conductive space saving kit consists of a 10 l space saving canister and a StopCock PA drain tap. The tap and canister can be grounded with the antistatic kit for use in potentially explosive atmospheres.
Catalog No. 50-000-25738 Supplier BURKLE INC Supplier No. 04350015
The electrically conductive space saving kit consists of a 10 l space saving canister and a StopCock PA drain tap. The tap and canister can be grounded with the antistatic kit for use in potentially explosive atmospheres.