Learn More
BURKLE INC Compact jerrycan & StopCock set, 1.32 gal. (5l), conductive
Catalog No. 5000025794
Encompass_Preferred
The electrically conductive space saving kit consists of a 5 l space saving canister and a StopCock PA drain tap. The tap and canister can be grounded with the antistatic kit for use in potentially explosive atmospheres.
Catalog No. 50-000-25794 Supplier BURKLE INC Supplier No. 04350005
The electrically conductive space saving kit consists of a 5 l space saving canister and a StopCock PA drain tap. The tap and canister can be grounded with the antistatic kit for use in potentially explosive atmospheres.