Learn More
Electron Microscopy Sciences Compact Low Cost Incubator, Model Imc18
SDP
Catalog No. 501902386
Encompass_Preferred
Low cost incubator for clinic or classroom uses cechanical convection technology. The easy-to-use interface has an internal light and window facilitate observation and LED display. Minimal benchtop for space restricted lab areas. Temperatures are accuracate. Applications include bacteriological cultures, hematology studies, and chick embryo studies.
Temperatures at or below ambient: 17° to 40°C
Footprint: 1.3 sq.ft (.12 sq. m).
Number of shelves (supplied/max.): 2/3.
Maximum shelf load: 4.4 lb. (2 kg).
Temperature uniformity of +/-1.2 degree C (at 37 degree C).
Temperature stability of +/- 0.2 degree C (at 37 degree C). One fixed shelf, shielded heating elements. Chamber volume: 19 L, dimensions: 12"W x 8"H x 12"D
Catalog No. 50-190-2386 Supplier Electron Microscopy Sciences Supplier No. 6315110
Low cost incubator for clinic or classroom uses cechanical convection technology. The easy-to-use interface has an internal light and window facilitate observation and LED display. Minimal benchtop for space restricted lab areas. Temperatures are accuracate. Applications include bacteriological cultures, hematology studies, and chick embryo studies.
Temperatures at or below ambient: 17° to 40°C
Footprint: 1.3 sq.ft (.12 sq. m).
Number of shelves (supplied/max.): 2/3.
Maximum shelf load: 4.4 lb. (2 kg).
Temperature uniformity of +/-1.2 degree C (at 37 degree C).
Temperature stability of +/- 0.2 degree C (at 37 degree C). One fixed shelf, shielded heating elements. Chamber volume: 19 L, dimensions: 12"W x 8"H x 12"D