Learn More
Research Products International Corp Compact Magnetic HotPlate Stirrer, Ceramic Plate Model
SDP
Catalog No. 501977192
Encompass
These Compact Magnetic stirrers are designed to provide years of rugged and dependable service in the laboratory environment.
Product Features:
Electronic speed control for constant speed even during changes in load
Low heat temperature stability
Ceramic steel plate for anti-corrosion and excellent thermal conductivity
Long life brushless motor
High magnetic adhesion
cTUVus approved
CE Marked
2 year warranty
Catalog No. 50-197-7192 Supplier Research Products International Corp Supplier No. 410038
These Compact Magnetic stirrers are designed to provide years of rugged and dependable service in the laboratory environment.
Product Features:
Electronic speed control for constant speed even during changes in load
Low heat temperature stability
Ceramic steel plate for anti-corrosion and excellent thermal conductivity
Long life brushless motor
High magnetic adhesion
cTUVus approved
CE Marked
2 year warranty