Learn More
Andwin Scientific Portable Photo Absorbance Sensor Battery Operated 3.6~4.5 Vdc
SDP
Catalog No. NC1769313
Encompass_Preferred
A photo absorbance sensor, weighing just over 200g, is a sleek, compact, and highly sensitive absorptiometer that enables efficient concentration measurement analysis. Built with intuitive bluetooth connectivity, the PiCOEXPLORER absorptiometer allows easy control of the unit from your mobile device for uninterrupted workflows and increased productivity. In addition, data management is improved with the seamless data transfer between PiCOEXPLORER and smartphone or tablet device.
Catalog No. NC1769313 Supplier Andwin Scientific Supplier No. PICOEXPLORER
A photo absorbance sensor, weighing just over 200g, is a sleek, compact, and highly sensitive absorptiometer that enables efficient concentration measurement analysis. Built with intuitive bluetooth connectivity, the PiCOEXPLORER absorptiometer allows easy control of the unit from your mobile device for uninterrupted workflows and increased productivity. In addition, data management is improved with the seamless data transfer between PiCOEXPLORER and smartphone or tablet device.