Learn More
Braintree Scientific Compact Portable Scale, 3000g Capacity, 1g Readability
SDP
Catalog No. NC1399123
Encompass
The CB is a compact portable scale which is ideally suited for a variety of uses and applications. It is small and easy to use. The bright green backlit LCD display shows clear results, with a simple to use keyboard and Grade 304 stainless steel removable top pan for easy cleaning. Weigh units: grams, ounces and pounds, selectable. The CB comes with batteries included. An optional mains adaptor can be purchased.Features Portable with battery power Stainless steel pan g, oz and lb Model Capacity Readability Platformsize CB-501 500g 0.1g 5.1? / 130mm CB-1001 1000g 0.1g 5.1? / 130mm CB-3000 3000g 1g 5.1? / 130mm **Sold for research purposes only and not for human use**
Catalog No. NC1399123 Supplier Braintree Scientific Supplier No. CB3000
The CB is a compact portable scale which is ideally suited for a variety of uses and applications. It is small and easy to use. The bright green backlit LCD display shows clear results, with a simple to use keyboard and Grade 304 stainless steel removable top pan for easy cleaning. Weigh units: grams, ounces and pounds, selectable. The CB comes with batteries included. An optional mains adaptor can be purchased.Features Portable with battery power Stainless steel pan g, oz and lb Model Capacity Readability Platformsize CB-501 500g 0.1g 5.1? / 130mm CB-1001 1000g 0.1g 5.1? / 130mm CB-3000 3000g 1g 5.1? / 130mm **Sold for research purposes only and not for human use**