Learn More
Felix Storch Inc ACCUCOLD COMPACT REFRIG SUMMIT SPRF26
SDP
Catalog No. NC0869259
Encompass_Preferred
Portable refrigerator or -18 C freezer, uses 12V (DC) or 24V (DC) or 110V (with AC adapter) power, digital thermostat; .88 cf, 17 x 21.5 x 14.25 (H x W xD): https://www.accucold.com/product/SPRF26
Catalog No. NC0869259 Supplier Felix Storch Inc Supplier No. SPRF26
Portable refrigerator or -18 C freezer, uses 12V (DC) or 24V (DC) or 110V (with AC adapter) power, digital thermostat; .88 cf, 17 x 21.5 x 14.25 (H x W xD): https://www.accucold.com/product/SPRF26