Learn More
BURKLE INC Compact spigot, cap nut 3/4" - NW xx mm
Catalog No. 5000025608
Encompass_Preferred
The spigot Compact for canisters and barrels captivates everyone through its compact build, which makes the valve on the canister ideal for use in refrigerators and on racks, etc. The short discharge does not protrude out over the lower edge of the container, which makes the valve a perfect choice even when space is very limited.
The discharge's specially optimised design creates an even stream of liquid with no splashing or spattering.
The spigot features easy motion and a tight seal. It is screwed on with a connecting cap nut. This guarantees a vertical position of the valve and makes it possible to adjust the valve later on.
PP, PE and PTFE are materials that offer outstanding chemical resistance to acids and alkalis.
Catalog No. 50-000-25608 Supplier BURKLE INC Supplier No. 05301000
The spigot Compact for canisters and barrels captivates everyone through its compact build, which makes the valve on the canister ideal for use in refrigerators and on racks, etc. The short discharge does not protrude out over the lower edge of the container, which makes the valve a perfect choice even when space is very limited.
The discharge's specially optimised design creates an even stream of liquid with no splashing or spattering.
The spigot features easy motion and a tight seal. It is screwed on with a connecting cap nut. This guarantees a vertical position of the valve and makes it possible to adjust the valve later on.
PP, PE and PTFE are materials that offer outstanding chemical resistance to acids and alkalis.