Learn More
Yamato Scientific America, Inc. COMPACT STERILIZER 18L 115V
Catalog No. NC2261587 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Economical, space-saving 18L compact steam sterilizer, ideal for research facilities. Easy to use. Powerful 2000W pipe heater. Easy to read 4 digit LED display. Programmable sterilizing and temperature functions. Timer Setting Range 0 to 999 min. Increased safety features: Water level detection sensor with alarm, Overheat Protection, Sensor Lid closure sensor (Inter lock), Pressure Lamp Indicator. Temp setting range 50 to 126°C
This item is not returnable. View return policy
Catalog No. NC2261587 Supplier Yamato Scientific America, Inc. Supplier No. SK102C
Economical, space-saving 18L compact steam sterilizer, ideal for research facilities. Easy to use. Powerful 2000W pipe heater. Easy to read 4 digit LED display. Programmable sterilizing and temperature functions. Timer Setting Range 0 to 999 min. Increased safety features: Water level detection sensor with alarm, Overheat Protection, Sensor Lid closure sensor (Inter lock), Pressure Lamp Indicator. Temp setting range 50 to 126°C