Learn More
Research Products International Corp Compact Utility Tub Cart, Three Shelf Unit, 18 x 24 x 39.25 Inches, Black
SDP
Catalog No. 501367656
Encompass
Chemical resistant utility carts a perfect for use in tight work areas where space is a premium. Ideal for storing or transporting laboratory reagents, equipment or other supplies. Utility carts feature a smooth surface for easy cleaning. Tub shelves safely stow items within 2.75 inch deep walls to prevent spills from reaching the floor. Constructed of heavy duty polypropylene for durability and high chemical resistance, cart easily bears the load in three sturdy shelves with a maximum weight of 300 pounds. Carts are impact resistant and will never rust, dent, scratch or stain. Swivel casters offer smooth transport and turning and two locking brakes.
Catalog No. 50-136-7656 Supplier Research Products International Corp Supplier No. 162704
Chemical resistant utility carts a perfect for use in tight work areas where space is a premium. Ideal for storing or transporting laboratory reagents, equipment or other supplies. Utility carts feature a smooth surface for easy cleaning. Tub shelves safely stow items within 2.75 inch deep walls to prevent spills from reaching the floor. Constructed of heavy duty polypropylene for durability and high chemical resistance, cart easily bears the load in three sturdy shelves with a maximum weight of 300 pounds. Carts are impact resistant and will never rust, dent, scratch or stain. Swivel casters offer smooth transport and turning and two locking brakes.