Learn More
Research Products International Corp Compact UV Lamp, 254 nm, 7 3/4 x 2 3/4 x 2 Inches
SDP
Catalog No. 50212695
Encompass
Easy to handle and convenient to use. Lightweight lamps feature a durable plastic housing. Shortwave model, 254 nm, is equipped with long-life filter.
Catalog No. 50-212-695 Supplier Research Products International Corp Supplier No. 353061
Add to Cart
Add to Cart
Easy to handle and convenient to use. Lightweight lamps feature a durable plastic housing. Shortwave model, 254 nm, is equipped with long-life filter.