Learn More
Grainger Siemens Sirius Compact Reversing Motor Starter, 3 Poles, Electronic Protection, 7 A, 1 ea
Catalog No. 502794918
Encompass_Preferred
This compact reversing motor starter is designed for the reliable control and protection of AC motors. Featuring integrated electronic overload protection and a 3-pole configuration, this module ensures efficient operation in a variety of industrial applications. It is engineered for DIN rail mounting and provides robust performance with vibration and shock resistance.
  • Reversing capability for versatile motor control
  • Electronic overload protection with a setting range of 1.6 A to 6.1 A
  • Integrated LED status indicator
  • 1NO/1NC auxiliary contacts included
  • Designed for 48 V to 500 V load voltage and 110 V to 230 V supply voltage
  • DIN rail mountable with screw termination
  • IP20 rated protection
  • Mechanical durability of 30,000,000 operations
Catalog No. 50-279-4918 Supplier Grainger Supplier No. 19YM58
This compact reversing motor starter is designed for the reliable control and protection of AC motors. Featuring integrated electronic overload protection and a 3-pole configuration, this module ensures efficient operation in a variety of industrial applications. It is engineered for DIN rail mounting and provides robust performance with vibration and shock resistance.
  • Reversing capability for versatile motor control
  • Electronic overload protection with a setting range of 1.6 A to 6.1 A
  • Integrated LED status indicator
  • 1NO/1NC auxiliary contacts included
  • Designed for 48 V to 500 V load voltage and 110 V to 230 V supply voltage
  • DIN rail mountable with screw termination
  • IP20 rated protection
  • Mechanical durability of 30,000,000 operations