Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New England Biolabs, Inc. COMPANION MODULEV2 24 REACTION
SDP

Catalog No. NC3256681
Encompass

  • Library preparation for ligation sequencing
  • Compatible with SQK-LSK114 protocols
  • Supports MinION, GridION, PromethION, and Flongle devices
  • Includes Salt-T4 DNA Ligase, NEBNext FFPE DNA Repair Buffer v2, NEBNext Ultra II Final Prep Enzyme Mix
  • 24 reactions
  • Storage temperature of -20°C
  • Sample type: DNA
  • Manual processing
  • Optimized reagent volumes
  • Simplified ordering and inventory management
  • No excess reagents or buffers

Catalog No. NC3256681 Supplier New England Biolabs, Inc. Supplier No. E7672S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Library preparation for ligation sequencing
  • Compatible with SQK-LSK114 protocols
  • Supports MinION, GridION, PromethION, and Flongle devices
  • Includes Salt-T4 DNA Ligase, NEBNext FFPE DNA Repair Buffer v2, NEBNext Ultra II Final Prep Enzyme Mix
  • 24 reactions
  • Storage temperature of -20°C
  • Sample type: DNA
  • Manual processing
  • Optimized reagent volumes
  • Simplified ordering and inventory management
  • No excess reagents or buffers

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.