Learn More
Bachem (Arg8)-Vasopressin (free acid) Trifluoroacetate
SDP
Supplier:  Bachem 4005475.0025
Encompass_Preferred
(Arg8)-Vasopressin (free acid) Trifluoroacetate, H-Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Arg-Gly-OH, 25255-33-8, C46H64N14O13S2, Mr 1085.23, 25mg. (Arg8,Gly-OH9)-Vasopressin, (Arg8)-Vasopressinoic acid, AVP (free acid), Argipressin (free acid), (Phe3,Arg8)-Oxytocin (free acid), Leiormone (free acid), Arginine Antidiuretic Hormone (free acid). Compared to AVP, AVP (free acid) shows markedly reduced antidiuretic activity and almost no vasopressor activity in vivo.
Catalog No. 50-194-7509