Learn More
Bachem Amyloid Beta Protein (1-40) Hydrochloride
SDP
Supplier:  Bachem 4038267.0100
Encompass_Preferred
Amyloid Beta Protein (1-40) Hydrochloride, H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH, 131438-79-4, C194H295N53O58S, Mr 4329.86, 0.1mg. Ab40 hydrochloride salt. Compared to the TFA salt, the HCl salt of amyloid b-protein (1-40) easily forms b-structures in PBS within a few hours at 25°C. b-structure formation is essential for amyloid peptide toxicity.
Catalog No. 50-194-6569