Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD CompBead Anti-Mouse Ig, κ/Negative Control Particles Set

Catalog No. bdb552843 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
6 mL Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552843 6 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552843 Supplier BD Supplier No. 552843
Only null left
Add to Cart
Add to Cart

Used to optimize fluorescence compensation settings for multicolor flow cytometric analyses. BD™ Anti-Mouse Ig, κ/Negative Control Particles Set includes two sets of polystyrene microparticles to distinctly stain positive and negative populations, especially when using tandem dye conjugates.

The set provides two populations of microparticles:

  • BD™ CompBeads Anti-Mouse Ig, κ—Binds any mouse κ light chain-bearing immunoglobulin
  • BD™ CompBeads Negative Control—Has no binding capacity

When mixed together with a fluorochrome-conjugated mouse antibody, the CompBeads provide distinct positive and negative (background fluorescence) stained populations which can be used to set compensation levels manually or using instrument set-up software.

Since the compensation adjustments are made using the same fluorochrome-labeled antibody to be used in the experiment, this method allows the investigator to more accurately establish compensation corrections for spectral overlap for any combination of fluorochrome-labeled antibodies (without having to use valuable tissue samples or hard-dyed beads with potentially mismatched fluorescence spectra).

Use of the BD™ CompBeads is highly recommended for use in all experiments using tandem dye (i.e., PE-Cy™7, APC-Cy™7, etc.) conjugates, which may have distinct spectral characteristics for each conjugate.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse Ig, κ
Applications Flow Cytometry
Conjugate Unconjugated
Formulation CompBead Anti-Mouse Ig, kappa Particles
CompBead Negative Control (FBS) Compensation Particles
Purification Method Unpurified
Quantity 6 mL Each
Primary or Secondary Primary
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.