Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Compbead anti-Rat Ig, κ/Negative Control (FBS) Compensation Particles Set, Clone: G16-510E3, BD

Catalog No. BDB552844 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
6 mL Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552844 6 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552844 Supplier BD Biosciences Supplier No. 552844
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The BD CompBeads Compensation Particles Anti-Rat Ig, kappa Set contains polystyrene microparticles, which are used to optimizefluorescence compensation settings for multicolor flow cytometric analyses. The set provides two populations of microparticles, the Anti-Rat Ig, kappa particles, which bind any rat kappa light chain-bearing immunoglobulin, and the Negative Control particles that have no binding capacity. When mixed together with a fluorochrome-conjugated rat antibody, the BD CompBeads Compensation Particles provide distinct positive and negative (background fluorescence) stained populations which can be used to set compensation levels manually or automatically using instrument set-up software. Since the compensation adjustments are made using the same fluorochrome-labeled antibody to be used in the experiment, this method allows the investigator to more accurately establish compensation corrections for spectral overlap for any combination of fluorochrome-labeled antibodies without having to use valuable tissue samples or hard-dyed beads with potentially mismatched fluorescence spectra. Use of the BD CompBeads Compensation Particles is highly recommended for use in all experiments using tandem dye (i.e., PE-Cy7, APC-Cy7, etc.) conjugates, which may have distinct spectral characteristics for each conjugate.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat Ig, κ
Classification Monoclonal
Conjugate Unconjugated
Purification Method Unpurified
Quantity 6 mL Each
Primary or Secondary Primary
Target Species Rat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.