Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Compbead Plus anti-Mouse Ig, κ/Negative Control (BSA) Compensation Plus (7.5μm) Particles Set, BD

Catalog No. BDB560497 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
6 mL Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560497 6 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560497 Supplier BD Biosciences Supplier No. 560497
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The BD CompBead Plus Anti-Mouse Ig, kappa beads are polystyrene particles used to establish fluorescence compensation settings for multicolor flow cytometric analyses. The kit provides two types of particles, the BD CompBead Plus Anti-Mouse Ig, kappa particles, which bind any mouse kappa light chain-bearing immunoglobulin, and the BD CompBead Plus Negative Control (BSA) particles, which have no binding capacity. When mixed together with a fluorochrome-conjugated mouse antibody, the BD CompBead Plus provides distinct positive and negative (background fluorescence) stained populations which can be used to set compensation levels manually or using instrument set-up software. Since the compensation adjustments are made using the same fluorochrome-labeled antibody to be used in the experiment, this method allows the investigator to more accurately establish compensation corrections for spectral overlap for any combination of fluorochrome-labeled antibodies (without having to use valuable tissue samples or hard-dyed beads with potentially mismatched fluorescence spectra). Use of the BD™ CompBead Plus is highly recommended for use in all experiments using tandem dye (i.e., PE-Cy7, APC-Cy7, etc.) conjugates, which may have distinct spectral characteristics for each conjugate.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig, κ
Conjugate Unconjugated
Purification Method Unpurified
Quantity 6 mL Each
Primary or Secondary Primary
Target Species Mouse
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.