Learn More
Apexbio Technology LLC EZ Cap EPO mRNA (ψUTP) 5x1mg (1mg/mL)
SDP
Catalog No. 502240944
Encompass_Preferred
EPO mRNA encodes human erythropoietin (EPO) protein a hormone that regulates erythropoiesis or red blood cell production EPO also plays roles in wound healing and the brain s response to neuronal injury Studies have shown that in vivo transfection with EPO mRNA leads to significant increases in reticulocyte count and hematocrit levels This mRNA utilizes co-transcriptional capping technology to incorporate a Cap 1 structure It is polyadenylated and incorporates UTP modification reducing the immune response in host cells and enhancing mRNA stability and translation efficiency
This item is not returnable. View return policy
Catalog No. 50-224-0944 Supplier Apexbio Technology LLC Supplier No. R10205
EPO mRNA encodes human erythropoietin (EPO) protein a hormone that regulates erythropoiesis or red blood cell production EPO also plays roles in wound healing and the brain s response to neuronal injury Studies have shown that in vivo transfection with EPO mRNA leads to significant increases in reticulocyte count and hematocrit levels This mRNA utilizes co-transcriptional capping technology to incorporate a Cap 1 structure It is polyadenylated and incorporates UTP modification reducing the immune response in host cells and enhancing mRNA stability and translation efficiency