Learn More
Biolegend COMPENSATION BEADS 100 TESTS
Catalog No. NC2181091
Encompass_Preferred
  • Designed for accurate and reliable fluorescence compensation.
  • Includes both positive and negative beads.
  • Enables precise spectral overlap correction.
  • Compatible with a wide range of species antibodies such as rabbit, donkey, hamster, human, rat, and mouse.
  • Features a ready-to-use solution format.
  • Compatible with traditional and spectral flow cytometers.
  • Conserves valuable experimental cells.
  • Provides uniform and bright staining.
  • Usage as compensation controls.
This item is not returnable. View return policy
Catalog No. NC2181091 Supplier Biolegend Supplier No. 424602
  • Designed for accurate and reliable fluorescence compensation.
  • Includes both positive and negative beads.
  • Enables precise spectral overlap correction.
  • Compatible with a wide range of species antibodies such as rabbit, donkey, hamster, human, rat, and mouse.
  • Features a ready-to-use solution format.
  • Compatible with traditional and spectral flow cytometers.
  • Conserves valuable experimental cells.
  • Provides uniform and bright staining.
  • Usage as compensation controls.