Learn More
Electron Microscopy Sciences Sure Shot Atomizer Compete Repair Kit
SDP
Catalog No. 5028846
Encompass_Preferred
Atomizer repair kit for Model B. Aluminum sprayers only. Contains: B05, B11, B12, B13, B25, 6 each of B15, B30, and B14.
Catalog No. 50-288-46 Supplier Electron Microscopy Sciences Supplier No. 64189
Atomizer repair kit for Model B. Aluminum sprayers only. Contains: B05, B11, B12, B13, B25, 6 each of B15, B30, and B14.