Learn More
Bachem p-Aminobenzamidine · 2 HCl
SDP
Supplier:  Bachem 4031123.0025
Encompass_Preferred
p-Aminobenzamidine · 2 HCl, 2498-50-2, C7H9N3 · 2 HCl, Mr 208.09, 25g. Competitive inhibitor of trypsin, plasmin and thrombin. CAS Number (net): 3858-83-1.
Catalog No. 50-194-6943