Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human COMP/TSP5 Protein, His Tag, Lyophilized, 1 mg

Catalog No. 501658028
Encompass_Preferred

Human COMP, also known as Thrombospondin-5 (TSP5), is a member of the thrombospondin family involved in maintaining cartilage structural integrity. This recombinant protein is expressed in human HEK293 cells, spanning Gln 21 to Ala 757, and features a C-terminal polyhistidine tag. It is supplied as a lyophilized powder formulated in 50 mM Tris, 150 mM NaCl, pH 7.5, with trehalose as a protectant.

  • Purity: ≥85% as determined by SDS-PAGE
  • Calculated molecular weight: 81.9 kDa (migrates as 90–100 kDa under reducing conditions due to glycosylation)
  • Endotoxin level: <0.1 EU per μg
  • Expressed from human HEK293 cells
  • Storage: Stable for 12 months at -20°C to -70°C in lyophilized state

Catalog No. 50-165-8028 Supplier ACROBiosystems Supplier No. COPH52201MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human COMP, also known as Thrombospondin-5 (TSP5), is a member of the thrombospondin family involved in maintaining cartilage structural integrity. This recombinant protein is expressed in human HEK293 cells, spanning Gln 21 to Ala 757, and features a C-terminal polyhistidine tag. It is supplied as a lyophilized powder formulated in 50 mM Tris, 150 mM NaCl, pH 7.5, with trehalose as a protectant.

  • Purity: ≥85% as determined by SDS-PAGE
  • Calculated molecular weight: 81.9 kDa (migrates as 90–100 kDa under reducing conditions due to glycosylation)
  • Endotoxin level: <0.1 EU per μg
  • Expressed from human HEK293 cells
  • Storage: Stable for 12 months at -20°C to -70°C in lyophilized state

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.