Learn More
Selleck Chemical LLC CompK-E1161-5MG
SDP
Catalog No. 503134438
Encompass_Preferred
CompK (compound K) a potent and selective hematopoietic progenitor kinase 1 (HPK1 or MAP4K1) small molecule inhibitor significantly improves human T-cell functions with enhanced T-cell receptor avidity to recognize tumor-associated antigens and tumor cytolytic activity by CD8 T cells
Catalog No. 50-313-4438 Supplier Selleck Chemical LLC Supplier No. E11615MG
Add to Cart
Add to Cart
CompK (compound K) a potent and selective hematopoietic progenitor kinase 1 (HPK1 or MAP4K1) small molecule inhibitor significantly improves human T-cell functions with enhanced T-cell receptor avidity to recognize tumor-associated antigens and tumor cytolytic activity by CD8 T cells