Learn More
Enzo Life Sciences Complement C3a des Arg (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900058
Encompass_Preferred
The Complement C3a des Arg (human), EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit comes ready-to-use with sample preparation reagents included. The Complement C3a des Arg (human), EIA kit is specific, offering low cross-reactivity to structurally related molecules, and sensitive with quantification of as little as 120 pg/mL Complement C3a des Arg. Sensitivity: 0.120 ng/ml (range 0.313 - 20 ng/ml). Use/Stability: Store all components at 4°C, except standard and conjugate at -20°C. UniProt ID: P01024. Assay Time: 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human . Content: GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer 10 concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Complement reagent A, Complement reagent B.
Catalog No. 50-200-6832