Learn More
ORS Nasco COMPL.FACEPIECE HEAD STRAP MED
Catalog No. 502551623
Encompass_Preferred
Honeywell part# RU65001M. Honeywell North RU6500 Series Full-Facepiece Air-Purifying Respirators Includes: Head Strap ANSI Standard: ANSI Z87.1 Black Full Facepiece Respirator Size: Medium Facepiece Material: Silicone For Use With: North N-Series line of cartridges filters and cartridge/filter combinations
Catalog No. 50-255-1623 Supplier ORS Nasco Supplier No. 068RU65001M
Honeywell part# RU65001M. Honeywell North RU6500 Series Full-Facepiece Air-Purifying Respirators Includes: Head Strap ANSI Standard: ANSI Z87.1 Black Full Facepiece Respirator Size: Medium Facepiece Material: Silicone For Use With: North N-Series line of cartridges filters and cartridge/filter combinations