Learn More
IBL America Complement System MBL Pathway
Catalog No. NC0837820
Encompass_Preferred
The Wieslab COMPL MP320 kit is an enzyme immunoassay for the determination of MBL complement pathway in human serum. For research use only, not for use in diagnostic procedures.
Catalog No. NC0837820 Supplier IBL America Supplier No. COMPL MP320 RUO
The Wieslab COMPL MP320 kit is an enzyme immunoassay for the determination of MBL complement pathway in human serum. For research use only, not for use in diagnostic procedures.