Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Complement C1s-A subcomponent (mouse) | >85.0% | 5 UG
SDP

Catalog No. 5000291334
Encompass_Preferred

Recombinant mouse C1s-A subcomponent is a serine protease of the classical complement pathway, expressed in mammalian cells and supplied as a filtered PBS solution for in vitro research applications. It is provided in small-quantity vials for biochemical and immunological assays.

  • Expressed in HEK293 cells.
  • C-terminal 10xHis tag for detection and purification.
  • Supplied as a 0.22 μm filtered PBS solution.
  • Purity greater than 85% by reducing SDS-PAGE.
  • Molecular weight approximately 76.6 kDa; glycosylated.
  • Endotoxin below 1 EU/μg as measured by LAL.
  • Store at -80°C and ship on dry ice; avoid repeated freeze-thaw cycles.

Catalog No. 50-002-91334 Supplier Medchemexpress LLC Supplier No. HYP754635UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse C1s-A subcomponent is a serine protease of the classical complement pathway, expressed in mammalian cells and supplied as a filtered PBS solution for in vitro research applications. It is provided in small-quantity vials for biochemical and immunological assays.

  • Expressed in HEK293 cells.
  • C-terminal 10xHis tag for detection and purification.
  • Supplied as a 0.22 μm filtered PBS solution.
  • Purity greater than 85% by reducing SDS-PAGE.
  • Molecular weight approximately 76.6 kDa; glycosylated.
  • Endotoxin below 1 EU/μg as measured by LAL.
  • Store at -80°C and ship on dry ice; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.