Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Complement C3/C3a B 20ug
SDP

Catalog No. 5000195845
Encompass_Preferred

Recombinant bovine complement C3a fragment supplied lyophilized for research applications in complement activation and immunology. The protein is expressed in E. coli with affinity tags to aid purification and is provided in a stabilized formulation suitable for storage and downstream assays.

  • Recombinant bovine C3a fragment expressed in E. coli for research use.
  • N-terminal His and SUMO tags facilitate purification and detection.
  • Lyophilized formulation with trehalose for improved stability during storage.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Observed molecular weight ~44 kDa under reducing conditions.
  • Supplied as 20 μg lyophilized aliquots; store at -20°C and freeze aliquots for long-term storage.

Catalog No. 50-001-95845 Supplier Medchemexpress LLC Supplier No. HYP7155220UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant bovine complement C3a fragment supplied lyophilized for research applications in complement activation and immunology. The protein is expressed in E. coli with affinity tags to aid purification and is provided in a stabilized formulation suitable for storage and downstream assays.

  • Recombinant bovine C3a fragment expressed in E. coli for research use.
  • N-terminal His and SUMO tags facilitate purification and detection.
  • Lyophilized formulation with trehalose for improved stability during storage.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Observed molecular weight ~44 kDa under reducing conditions.
  • Supplied as 20 μg lyophilized aliquots; store at -20°C and freeze aliquots for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.