Learn More
Medchemexpress LLC Complement C3/C3a H 100ug | 100ug
SDP
Catalog No. 5000194487
Encompass_Preferred
Complement C3/C3a protein plays a key role in initiating the complement system through processing by C3 convertase in both the classical and alternative pathways Complement C3/C3a protein is involved in various immune and inflammatory responses and also has a role in regulating blood pressure Complement C3/C3a Protein Human is a recombinant Complement C3/C3a protein expressed in E coli untagged and consists of 77 amino acids (S672-R748)[1][2][3][4][5][6][7][8][9][10][11]
Catalog No. 50-001-94487 Supplier Medchemexpress LLC Supplier No. HYP7862100UG
Complement C3/C3a protein plays a key role in initiating the complement system through processing by C3 convertase in both the classical and alternative pathways Complement C3/C3a protein is involved in various immune and inflammatory responses and also has a role in regulating blood pressure Complement C3/C3a Protein Human is a recombinant Complement C3/C3a protein expressed in E coli untagged and consists of 77 amino acids (S672-R748)[1][2][3][4][5][6][7][8][9][10][11]