Learn More
Medchemexpress LLC Complement C3/C3a M 1mg | 1mg
SDP
Catalog No. 5000194115
Encompass_Preferred
Complement C3/C3a protein activates the complement system playing a central role in both classical and alternative pathways C3b binds covalently to cell surface carbohydrates or immune aggregates while C3a acts as an inflammatory mediator inducing neutrophil chemoattraction and promoting smooth muscle contraction increased vascular permeability and histamine release The shorter isoform of C3a stimulates B-cells Complement C3/C3a Protein Mouse is the recombinant mouse-derived Complement C3/C3a protein expressed by E coli with tag free
Catalog No. 50-001-94115 Supplier Medchemexpress LLC Supplier No. HYP78631MG
Complement C3/C3a protein activates the complement system playing a central role in both classical and alternative pathways C3b binds covalently to cell surface carbohydrates or immune aggregates while C3a acts as an inflammatory mediator inducing neutrophil chemoattraction and promoting smooth muscle contraction increased vascular permeability and histamine release The shorter isoform of C3a stimulates B-cells Complement C3/C3a Protein Mouse is the recombinant mouse-derived Complement C3/C3a protein expressed by E coli with tag free