Learn More
Athens Research And Technology Inc COMPLEMENT C3 HU 1MG
Supplier:  Athens Research And Technology Inc 16160303031MG
Encompass_Preferred
Complement C3 Human Plasma: 1 MG; Frozen in PBS, pH 7.4; Purity: > =95% by SDS-PAGE. Reacts to C3 antisera on Western Blot. Negative against C4 antisera on Western Blot. This native protein is prepared from human plasma in Athens Research & Tech lab.
Catalog No. 50-489-718