Learn More
Medchemexpress LLC Complement C3 protein, human (HEK293, His) | >95.0% | 500UG
SDP
Catalog No. 5000174357
Encompass_Preferred
Recombinant human complement C3 protein produced in HEK293 cells with a C-terminal His tag. Supplied as a lyophilized, glycosylated protein (sequence S23-N1663) for research use in complement activation, binding studies, and assay development. Purity >95% by reducing SDS-PAGE; multiple apparent molecular weights reflect glycosylation.
  • Glycosylated recombinant human C3 spanning S23-N1663.
  • Expressed in HEK293 cells for humanlike post-translational modifications.
  • C-terminal His tag enables affinity purification and detection.
  • >95.0% purity by reducing SDS-PAGE.
  • Lyophilized from PBS with 8% trehalose for storage stability.
  • Multiple apparent molecular weights due to glycosylation.
  • Suitable for complement activation assays and binding studies.
  • Recommended storage at -20°C; aliquot and freeze after reconstitution for extended storage.
Catalog No. 50-001-74357 Supplier Medchemexpress LLC Supplier No. HYP702819500UG
Recombinant human complement C3 protein produced in HEK293 cells with a C-terminal His tag. Supplied as a lyophilized, glycosylated protein (sequence S23-N1663) for research use in complement activation, binding studies, and assay development. Purity >95% by reducing SDS-PAGE; multiple apparent molecular weights reflect glycosylation.
  • Glycosylated recombinant human C3 spanning S23-N1663.
  • Expressed in HEK293 cells for humanlike post-translational modifications.
  • C-terminal His tag enables affinity purification and detection.
  • >95.0% purity by reducing SDS-PAGE.
  • Lyophilized from PBS with 8% trehalose for storage stability.
  • Multiple apparent molecular weights due to glycosylation.
  • Suitable for complement activation assays and binding studies.
  • Recommended storage at -20°C; aliquot and freeze after reconstitution for extended storage.