Learn More
Medchemexpress LLC Complement C3 Mouse 50ug | 50ug
SDP
Catalog No. 5000193597
Encompass_Preferred
Complement C3 protein activates the complement system playing a central role in both classical and alternative pathways C3b binds covalently to cell surface carbohydrates or immune aggregates while C3a acts as an inflammatory mediator inducing neutrophil chemoattraction and promoting smooth muscle contraction increased vascular permeability and histamine release The shorter isoform of C3a stimulates B-cells Complement C3 Protein Mouse (HEK293 His) is the recombinant mouse-derived Complement C3 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-93597 Supplier Medchemexpress LLC Supplier No. HYP7824750UG
Complement C3 protein activates the complement system playing a central role in both classical and alternative pathways C3b binds covalently to cell surface carbohydrates or immune aggregates while C3a acts as an inflammatory mediator inducing neutrophil chemoattraction and promoting smooth muscle contraction increased vascular permeability and histamine release The shorter isoform of C3a stimulates B-cells Complement C3 Protein Mouse (HEK293 His) is the recombinant mouse-derived Complement C3 protein expressed by HEK293 with C-His labeled tag