Learn More
Medchemexpress LLC Complement C3 protein, cynomolgus (HEK293, His) | >90.0% | 5 UG
SDP
Catalog No. 5000291062
Encompass_Preferred
Recombinant complement C3 protein derived from cynomolgus macaque and expressed in HEK293 cells with a C-terminal His tag. It is suitable for studies of complement activation, C3 processing, and complement-mediated inflammation, and for use as a reagent in biochemical and cell-based assays.
  • Cynomolgus origin for macaque-specific studies.
  • Expressed in HEK293 cells to retain mammalian post-translational modifications.
  • C-terminal His tag enables purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Available in small research sizes for assay development.
Catalog No. 50-002-91062 Supplier Medchemexpress LLC Supplier No. HYP753325UG
Recombinant complement C3 protein derived from cynomolgus macaque and expressed in HEK293 cells with a C-terminal His tag. It is suitable for studies of complement activation, C3 processing, and complement-mediated inflammation, and for use as a reagent in biochemical and cell-based assays.
  • Cynomolgus origin for macaque-specific studies.
  • Expressed in HEK293 cells to retain mammalian post-translational modifications.
  • C-terminal His tag enables purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Available in small research sizes for assay development.