Learn More
Alpco Ltd Complement C4d ELISA
SDP
Supplier:  Alpco Ltd 13COMPLC4D
Encompass
Complement C4d is a highly specific complement activity marker for the classical and lectin pathway. The C4d ELISA is highly-specific and reliable as it targets a unique neoepitope only present in the C4d fragment, which enables specific assessment of complement activation and does not measure native C4 or other C4-fragments. The Complement System C4d assay includes ready-to-use reagents and offers short incubation times, reducing hands-on time and simplifying workflow. As an early activity marker in the classical and lectin pathways, C4d can help in evaluation of complement therapeutics targeting the upstream steps of the activation cascade.
Catalog No. NC2284548