Learn More
Medchemexpress LLC Complement C5/C5a C 100ug | 100ug
SDP
Catalog No. 5000194101
Encompass_Preferred
Complement C5 is activated by C5 convertase inducing C5-C9 assembly to form the membrane attack complex The transient C6 binding site on C5b is critical for the formation of the cleavage complex Complement C5/C5a Protein Cynomolgus is the recombinant cynomolgus-derived Complement C5/C5a protein expressed by E coli with tag free
Catalog No. 50-001-94101 Supplier Medchemexpress LLC Supplier No. HYP78595100UG
Complement C5 is activated by C5 convertase inducing C5-C9 assembly to form the membrane attack complex The transient C6 binding site on C5b is critical for the formation of the cleavage complex Complement C5/C5a Protein Cynomolgus is the recombinant cynomolgus-derived Complement C5/C5a protein expressed by E coli with tag free