Learn More
Medchemexpress LLC Complement C5/C5a H 5ug | 5ug
SDP
Catalog No. 5000194483
Encompass_Preferred
Complement C5/C5a Protein Human is a recombinant human complement C5a C5a is a 74-amino acid glycoprotein generated on activation of the C system Complement C5 is cleaved by proteolysis in the terminal phase of complement activation generating the pro-inflammatory C5a and membrane attack complex nucleator C5b C5a is an inflammatory mediator generated by complement activation that positively regulates various arms of immune defense including Toll-like receptor 4 (TLR4) signaling Complement C5/C5a Protein Human is a human-derived recombinant protein that expressed by E coli[1][2]
Catalog No. 50-001-94483 Supplier Medchemexpress LLC Supplier No. HYP78645UG
Complement C5/C5a Protein Human is a recombinant human complement C5a C5a is a 74-amino acid glycoprotein generated on activation of the C system Complement C5 is cleaved by proteolysis in the terminal phase of complement activation generating the pro-inflammatory C5a and membrane attack complex nucleator C5b C5a is an inflammatory mediator generated by complement activation that positively regulates various arms of immune defense including Toll-like receptor 4 (TLR4) signaling Complement C5/C5a Protein Human is a human-derived recombinant protein that expressed by E coli[1][2]