Learn More
Medchemexpress LLC Complement C5/C5a M 5ug | 5ug
SDP
Catalog No. 5000185737
Encompass_Preferred
Complement C5/C5a Protein Mouse is a recombinant mouse complement component 5a (C5a) C5a is a potent pro-inflammatory mediator and acts as an essential part of the innate immune response[1]
Catalog No. 50-001-85737 Supplier Medchemexpress LLC Supplier No. HYP7695A5UG
Complement C5/C5a Protein Mouse is a recombinant mouse complement component 5a (C5a) C5a is a potent pro-inflammatory mediator and acts as an essential part of the innate immune response[1]