Learn More
Medchemexpress LLC Complement C5/C5a protein, human (HEK293, His) | >95.0% | 100UG
SDP
Catalog No. 5000175723
Encompass_Preferred
Recombinant human Complement C5 produced in HEK293 cells with a C-terminal His tag. C5 is cleaved into C5a and C5b during complement activation and contributes to membrane attack complex formation. Supplied lyophilized from PBS with trehalose, this protein is intended for biochemical and immunological assays.
Catalog No. 50-001-75723 Supplier Medchemexpress LLC Supplier No. HYP700862100UG
Recombinant human Complement C5 produced in HEK293 cells with a C-terminal His tag. C5 is cleaved into C5a and C5b during complement activation and contributes to membrane attack complex formation. Supplied lyophilized from PBS with trehalose, this protein is intended for biochemical and immunological assays.